missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzaldehyde 98.0+%, TCI America™

Catalog No. H019825G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H019825G 25 g
H0198100G 100 g
H0198500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H019825G Supplier TCI America Supplier No. H019825G

Chemical Identifiers

CAS 123-08-0
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00006939
InChI Key RGHHSNMVTDWUBI-UHFFFAOYSA-N
Synonym p-hydroxybenzaldehyde, 4-formylphenol, p-formylphenol, p-oxybenzaldehyde, benzaldehyde, 4-hydroxy, parahydroxybenzaldehyde, benzaldehyde, p-hydroxy, 4-hydroxy-benzaldehyde, usaf m-6, 4-hydroxy benzaldehyde
PubChem CID 126
ChEBI CHEBI:17597
IUPAC Name 4-hydroxybenzaldehyde
SMILES C1=CC(=CC=C1C=O)O

Specifications

Melting Point 117°C
Quantity 25 g
Formula Weight 122.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxybenzaldehyde
EINECSNumber (3)-1183&(3)-2660
RTECSNumber CU6475000
TSCA Yes