Learn More
4-Hydroxybenzaldehyde, 99%
CAS: 123-08-0 | C7H6O2 | 122.12 g/mol
Supplier: Thermo Scientific Chemicals 162770010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Hydroxybenzaldehyde | |
| 114.0°C to 118.0°C | |
| Authentic | |
| Plastic bottle | |
| HOC6H4CHO | |
| 08,64 | |
| p-hydroxybenzaldehyde, 4-formylphenol, p-formylphenol, p-oxybenzaldehyde, benzaldehyde, 4-hydroxy, parahydroxybenzaldehyde, benzaldehyde, p-hydroxy, 4-hydroxy-benzaldehyde, usaf m-6, 4-hydroxy benzaldehyde | |
| RGHHSNMVTDWUBI-UHFFFAOYSA-N | |
| 4-hydroxybenzaldehyde | |
| 126 | |
| 122.12 | |
| Crystalline Powder |
| 123-08-0 | |
| Yellow to Brown | |
| 98.5% min. (GC) | |
| C7H6O2 | |
| 1 kg | |
| 15,4849 | |
| Solubility in water: 13g/L (30°C). Other solubilities: 36.8g/L benzene (65°C) | |
| C1=CC(=CC=C1C=O)O | |
| 122.12 | |
| CHEBI:17597 | |
| 99% |
Chemical Identifiers
| 123-08-0 | |
| 122.12 | |
| p-hydroxybenzaldehyde, 4-formylphenol, p-formylphenol, p-oxybenzaldehyde, benzaldehyde, 4-hydroxy, parahydroxybenzaldehyde, benzaldehyde, p-hydroxy, 4-hydroxy-benzaldehyde, usaf m-6, 4-hydroxy benzaldehyde | |
| CHEBI:17597 | |
| C1=CC(=CC=C1C=O)O |
| C7H6O2 | |
| RGHHSNMVTDWUBI-UHFFFAOYSA-N | |
| 126 | |
| 4-hydroxybenzaldehyde |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec
GHS Signal Word: Warning
EINECSNumber : 204-599-1
RUO – Research Use Only