missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzaldehyde, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 162770025

Catalog No. AC162770025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-Hydroxybenzaldehyde
Authentic
Plastic bottle
HOC6H4CHO
08,64
p-hydroxybenzaldehyde, 4-formylphenol, p-formylphenol, p-oxybenzaldehyde, benzaldehyde, 4-hydroxy, parahydroxybenzaldehyde, benzaldehyde, p-hydroxy, 4-hydroxy-benzaldehyde, usaf m-6, 4-hydroxy benzaldehyde
RGHHSNMVTDWUBI-UHFFFAOYSA-N
4-hydroxybenzaldehyde
126
122.12
123-08-0
98.5% min. (GC)
C7H6O2
2.5kg
15,4849
Solubility in water: 13g/L (30°C). Other solubilities: 36.8g/L benzene (65°C)
C1=CC(=CC=C1C=O)O
122.12
CHEBI:17597
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec

GHS Signal Word: Warning

EINECSNumber : 204-599-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only