Learn More
4-Hydroxybenzaldehyde, 98%
CAS: 123-08-0 | C7H6O2 | 122.123 g/mol
Supplier: Thermo Scientific Chemicals A1358022
Description
4-Hydroxybenzaldehyde is used as a chemical that maintains bactericidal activity when tested against certain bacteria strains.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Hydroxybenzaldehyde is used as a chemical that maintains bactericidal activity when tested against certain bacteria strains.
Solubility
Soluble in water(13.8g/L), ether, alcohol, and acetone.
Notes
Air Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
| 4-Hydroxybenzaldehyde | |
| 115°C to 119°C | |
| 191°C (50 mmHg) | |
| C7H6O2 | |
| 100 g | |
| Air Sensitive | |
| p-hydroxybenzaldehyde, 4-formylphenol, p-formylphenol, p-oxybenzaldehyde, benzaldehyde, 4-hydroxy, parahydroxybenzaldehyde, benzaldehyde, p-hydroxy, 4-hydroxy-benzaldehyde, usaf m-6, 4-hydroxy benzaldehyde | |
| RGHHSNMVTDWUBI-UHFFFAOYSA-N | |
| 4-hydroxybenzaldehyde | |
| 126 | |
| 122.12 |
| 123-08-0 | |
| 1.129 | |
| 174°C (345°F) | |
| MFCD00006939 | |
| 471352 | |
| 14,4811 | |
| Soluble in water(13.8g/L),ether,alcohol,and acetone. | |
| C1=CC(=CC=C1C=O)O | |
| 122.123 | |
| CHEBI:17597 | |
| 98% |
Chemical Identifiers
| 123-08-0 | |
| 122.123 | |
| RGHHSNMVTDWUBI-UHFFFAOYSA-N | |
| 126 | |
| 4-hydroxybenzaldehyde |
| C7H6O2 | |
| MFCD00006939 | |
| p-hydroxybenzaldehyde, 4-formylphenol, p-formylphenol, p-oxybenzaldehyde, benzaldehyde, 4-hydroxy, parahydroxybenzaldehyde, benzaldehyde, p-hydroxy, 4-hydroxy-benzaldehyde, usaf m-6, 4-hydroxy benzaldehyde | |
| CHEBI:17597 | |
| C1=CC(=CC=C1C=O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 204-599-1
RTECSNumber : CU6475000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only