missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzaldehyde, 98%

Catalog No. AAA1358014
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1358014 25 g
AAA1358022 100 g
AAA1358036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1358014 Supplier Thermo Scientific Chemicals Supplier No. A1358014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 123-08-0 | C7H6O2 | 122.123 g/mol

4-Hydroxybenzaldehyde is used as a chemical that maintains bactericidal activity when tested against certain bacteria strains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzaldehyde is used as a chemical that maintains bactericidal activity when tested against certain bacteria strains.

Solubility
Soluble in water(13.8g/L), ether, alcohol, and acetone.

Notes
Air Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 123-08-0
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00006939
InChI Key RGHHSNMVTDWUBI-UHFFFAOYSA-N
Synonym p-hydroxybenzaldehyde, 4-formylphenol, p-formylphenol, p-oxybenzaldehyde, benzaldehyde, 4-hydroxy, parahydroxybenzaldehyde, benzaldehyde, p-hydroxy, 4-hydroxy-benzaldehyde, usaf m-6, 4-hydroxy benzaldehyde
PubChem CID 126
ChEBI CHEBI:17597
IUPAC Name 4-hydroxybenzaldehyde
SMILES C1=CC(=CC=C1C=O)O

Specifications

Melting Point 115°C to 119°C
Density 1.129
Boiling Point 191°C (50 mmHg)
Flash Point 174°C (345°F)
Quantity 25 g
Beilstein 471352
Sensitivity Air Sensitive
Merck Index 14,4811
Solubility Information Soluble in water(13.8g/L),ether,alcohol,and acetone.
Formula Weight 122.12
Percent Purity 98%
Chemical Name or Material 4-Hydroxybenzaldehyde
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 204-599-1
RTECSNumber CU6475000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only