missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Hydroxyazobenzene-2-carboxylic Acid 98.0+%, TCI America™
Supplier: TCI America H058625G
Specifications
| 4′-Hydroxyazobenzene-2-carboxylic Acid | |
| 201°C | |
| MFCD00002428 | |
| 2-4-hydroxyphenylazo benzoic acid, haba, 2-p-hydroxyphenylazo benzoic acid, benzoic acid, 2-4-hydroxyphenyl azo, o-p-hydroxyphenylazo benzoic acid, 2-4'-hydroxyphenyl-azo benzoic acid, hbaba, 2-4-hydroxyphenyl diazenyl benzoic acid, 2-2-4-hydroxyphenyl diazen-1-yl benzoic acid, benzoic acid, o-p-hydroxyphenyl azo | |
| C1=CC=C(C(=C1)C(=O)O)NN=C2C=CC(=O)C=C2 | |
| 242.234 | |
| 242.23 | |
| Crystalline Powder |
| 1634-82-8 | |
| C13H10N2O3 | |
| 25 g | |
| FBVSMDPNVYJNON-UHFFFAOYSA-N | |
| 2-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzoic acid | |
| 5357439 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 1634-82-8 | |
| 242.234 | |
| FBVSMDPNVYJNON-UHFFFAOYSA-N | |
| 5357439 | |
| C1=CC=C(C(=C1)C(=O)O)NN=C2C=CC(=O)C=C2 |
| C13H10N2O3 | |
| MFCD00002428 | |
| 2-4-hydroxyphenylazo benzoic acid, haba, 2-p-hydroxyphenylazo benzoic acid, benzoic acid, 2-4-hydroxyphenyl azo, o-p-hydroxyphenylazo benzoic acid, 2-4'-hydroxyphenyl-azo benzoic acid, hbaba, 2-4-hydroxyphenyl diazenyl benzoic acid, 2-2-4-hydroxyphenyl diazen-1-yl benzoic acid, benzoic acid, o-p-hydroxyphenyl azo | |
| 2-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzoic acid |
Safety and Handling
TSCA : Yes