missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxyazobenzene-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. H05865G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05865G 5 g
H058625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05865G Supplier TCI America Supplier No. H05865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1634-82-8
Molecular Formula C13H10N2O3
Molecular Weight (g/mol) 242.234
MDL Number MFCD00002428
InChI Key FBVSMDPNVYJNON-UHFFFAOYSA-N
Synonym 2-4-hydroxyphenylazo benzoic acid, haba, 2-p-hydroxyphenylazo benzoic acid, benzoic acid, 2-4-hydroxyphenyl azo, o-p-hydroxyphenylazo benzoic acid, 2-4'-hydroxyphenyl-azo benzoic acid, hbaba, 2-4-hydroxyphenyl diazenyl benzoic acid, 2-2-4-hydroxyphenyl diazen-1-yl benzoic acid, benzoic acid, o-p-hydroxyphenyl azo
PubChem CID 5357439
IUPAC Name 2-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)NN=C2C=CC(=O)C=C2

Specifications

Melting Point 201°C
Quantity 5 g
Formula Weight 242.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Hydroxyazobenzene-2-carboxylic Acid