missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxyacetophenone 98.0+%, TCI America™

Catalog No. H0193500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0193500G 500 g
H019325G 25 g
H0193100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0193500G Supplier TCI America Supplier No. H0193500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99-93-4
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002359
InChI Key TXFPEBPIARQUIG-UHFFFAOYSA-N
Synonym 4'-hydroxyacetophenone, 4-hydroxyacetophenone, 1-4-hydroxyphenyl ethanone, 4-acetylphenol, p-hydroxyacetophenone, piceol, ethanone, 1-4-hydroxyphenyl, p-acetylphenol, para-hydroxyacetophenone, p-oxyacetophenone
PubChem CID 7469
ChEBI CHEBI:28032
IUPAC Name 1-(4-hydroxyphenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 110°C
Color White
Boiling Point 148°C
Quantity 500 g
Formula Weight 136.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Hydroxyacetophenone
EINECSNumber (3)-4119
RTECSNumber AM8750000
TSCA Yes