missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxyacetophenone, 99%

CAS: 99-93-4 | C8H8O2 | 136.15 g/mol

Supplier:  Thermo Scientific Chemicals A119740B

 View more versions of this product

Catalog No. AAA119740B


Only null left
Explore available promotions

Description

Description

4?-Hydroxyacetophenone has been used as ketone component in the preparation of 1-aryl-3-phenethylamino-1-propanone hydrochlorides, potential cytotoxic agents, via Mannich reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Hydroxyacetophenone has been used as ketone component in the preparation of 1-aryl-3-phenethylamino-1-propanone hydrochlorides, potential cytotoxic agents, via Mannich reactions.

Solubility
Soluble in 95% Ethanol. Slightly soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications

Specifications

4'-Hydroxyacetophenone
106°C to 108°C
166°C (330°F)
MFCD00002359
774355
Soluble in 95% Ethanol. Slightly soluble in water.
CC(=O)C1=CC=C(O)C=C1
136.15
CHEBI:28032
99%
99-93-4
286°C
C8H8O2
1000 g
4'-hydroxyacetophenone, 4-hydroxyacetophenone, 1-4-hydroxyphenyl ethanone, 4-acetylphenol, p-hydroxyacetophenone, piceol, ethanone, 1-4-hydroxyphenyl, p-acetylphenol, para-hydroxyacetophenone, p-oxyacetophenone
TXFPEBPIARQUIG-UHFFFAOYSA-N
1-(4-hydroxyphenyl)ethanone
7469
136.15

Chemical Identifiers

99-93-4
136.15
TXFPEBPIARQUIG-UHFFFAOYSA-N
7469
1-(4-hydroxyphenyl)ethanone
C8H8O2
MFCD00002359
4'-hydroxyacetophenone, 4-hydroxyacetophenone, 1-4-hydroxyphenyl ethanone, 4-acetylphenol, p-hydroxyacetophenone, piceol, ethanone, 1-4-hydroxyphenyl, p-acetylphenol, para-hydroxyacetophenone, p-oxyacetophenone
CHEBI:28032
CC(=O)C1=CC=C(O)C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 202-802-8

RTECSNumber : AM8750000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only