missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxyacetophenone, 99%

Catalog No. AAA1197436
Change view
Click to view available options
Quantity:
100 g
500 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1197436 500 g
AAA1197422 100 g
AAA119740B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1197436 Supplier Thermo Scientific Chemicals Supplier No. A1197436
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99-93-4 | C8H8O2 | 136.15 g/mol

4?-Hydroxyacetophenone has been used as ketone component in the preparation of 1-aryl-3-phenethylamino-1-propanone hydrochlorides, potential cytotoxic agents, via Mannich reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Hydroxyacetophenone has been used as ketone component in the preparation of 1-aryl-3-phenethylamino-1-propanone hydrochlorides, potential cytotoxic agents, via Mannich reactions.

Solubility
Soluble in 95% Ethanol. Slightly soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 99-93-4
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002359
InChI Key TXFPEBPIARQUIG-UHFFFAOYSA-N
Synonym 4'-hydroxyacetophenone, 4-hydroxyacetophenone, 1-4-hydroxyphenyl ethanone, 4-acetylphenol, p-hydroxyacetophenone, piceol, ethanone, 1-4-hydroxyphenyl, p-acetylphenol, para-hydroxyacetophenone, p-oxyacetophenone
PubChem CID 7469
ChEBI CHEBI:28032
IUPAC Name 1-(4-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 106°C to 108°C
Boiling Point 286°C
Flash Point 166°C (330°F)
Quantity 500 g
Beilstein 774355
Solubility Information Soluble in 95% Ethanol. Slightly soluble in water.
Formula Weight 136.15
Percent Purity 99%
Chemical Name or Material 4'-Hydroxyacetophenone
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 202-802-8
RTECSNumber AM8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only