missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-N-methylpiperidine, 98%, Thermo Scientific™

Catalog No. p-146171
Change view
Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC121510050 5g Glass bottle
AC121511000 100g Glass bottle
AC121515000 500g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC121510050 Supplier Thermo Scientific Chemicals Supplier No. 121510050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 106-52-5
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.17
MDL Number MFCD00006500
InChI Key BAUWRHPMUVYFOD-UHFFFAOYSA-N
Synonym 4-hydroxy-1-methylpiperidine, n-methyl-4-piperidinol, 1-methyl-4-piperidinol, 4-hydroxy-n-methylpiperidine, n-methyl-4-hydroxypiperidine, 4-piperidinol, 1-methyl, 1-methyl-4-hydroxypiperidine, n-methylpiperidol, n-methyl-4-hydroxy piperidine, 4-hydroxy-1-methyl-piperidine
PubChem CID 66048
IUPAC Name 1-methylpiperidin-4-ol
SMILES CN1CCC(CC1)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Boiling Point 200°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4760 to 1.479
Quantity 5g
Beilstein 21, I, 188
Solubility Information Solubility in water: miscible with water
Formula Weight 115.17
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-N-methylpiperidine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-406-8
TSCA TSCA

RUO – Research Use Only