missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-6-methylcoumarin 98.0+%, TCI America™

Catalog No. H14215G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14215G 5 g
H14211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14215G Supplier TCI America Supplier No. H14215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13252-83-0
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.17
MDL Number MFCD00085738
InChI Key CSVFZDYJGBVNEX-UHFFFAOYSA-N
Synonym 4-hydroxy-6-methylcoumarin, 4-hydroxy-6-methyl-2h-chromen-2-one, 6-methyl-4-hydroxycoumarin, 2h-1-benzopyran-2-one,4-hydroxy-6-methyl, 2h-1-benzopyran-2-one, 4-hydroxy-6-methyl, 2-hydroxy-6-methylchromen-4-one, 2-hydroxy-6-methyl-4h-chromen-4-one, 6-methyl-4-hydroxy coumarin, hydroxy-6-methylcoumarin,4
PubChem CID 54686349
IUPAC Name 2-hydroxy-6-methyl-4H-chromen-4-one
SMILES CC1=CC=C2OC(O)=CC(=O)C2=C1

Specifications

Melting Point 261°C
Color White
Quantity 5 g
Formula Weight 176.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-6-methylcoumarin