missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-6-methyl-2-pyrone 98.0+%, TCI America™

Catalog No. p-7130684
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
H071525G 25 g
H0715500G 500 g
2 options

Catalog No. H071525G

Supplier: TCI America H071525G

Only null left

Chemical Identifiers

CAS 675-10-5
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00006641,MFCD18820248
InChI Key OOKCZXGEYPSNIM-UHFFFAOYSA-N
Synonym 4-hydroxy-6-methyl-2-pyrone, 2-hydroxy-6-methyl-4h-pyran-4-one, 4-hydroxy-6-methyl-2h-pyran-2-one, triacetic acid lactone, triacetate lactone, 2h-pyran-2-one, 4-hydroxy-6-methyl, 6-methyl-4-hydroxypyron-2, unii-s1s883s4ee, 6-methyl-4-hydroxy-2-pyrone, ccris 3600
PubChem CID 54675757
ChEBI CHEBI:16458
IUPAC Name 2-hydroxy-6-methyl-4H-pyran-4-one
SMILES CC1=CC(=O)C=C(O)O1

Specifications

Melting Point 188°C
Quantity 25 g
Formula Weight 126.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-6-methyl-2-pyrone
RTECSNumber UQ1000000
TSCA Yes