missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-6-methyl-2-pyrone, 98%

Catalog No. p-7045641
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1145714 25 g
AAL1145722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1145714 Supplier Thermo Scientific Chemicals Supplier No. L1145714

CAS: 675-10-5 | C6H6O3 | 126.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 675-10-5
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00006641,MFCD18820248
InChI Key OOKCZXGEYPSNIM-UHFFFAOYSA-N
Synonym 4-hydroxy-6-methyl-2-pyrone, 2-hydroxy-6-methyl-4h-pyran-4-one, 4-hydroxy-6-methyl-2h-pyran-2-one, triacetic acid lactone, triacetate lactone, 2h-pyran-2-one, 4-hydroxy-6-methyl, 6-methyl-4-hydroxypyron-2, unii-s1s883s4ee, 6-methyl-4-hydroxy-2-pyrone, ccris 3600
PubChem CID 54675757
ChEBI CHEBI:16458
IUPAC Name 4-hydroxy-6-methylpyran-2-one
SMILES CC1=CC(=O)C=C(O)O1

Specifications

Melting Point ∽186°C (decomposition)
Quantity 25 g
Beilstein 113815
Formula Weight 126.11
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-6-methyl-2-pyrone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-619-2
RTECSNumber UQ1000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only