missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-5-iodopyrimidine, 95%, Thermo Scientific Chemicals

$273.62 - $273.62

Chemical Identifiers

CAS 4349-07-9
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6119003
View Documents
Thermo Scientific Chemicals
H6119003
1 g
Each for $273.62
Only null left
 
Description

Description

4-Hydroxy-5-iodopyrimidine is an intermediate used in pharmaceuticals and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-5-iodopyrimidine is an intermediate used in pharmaceuticals and chemical research.

Solubility
Soluble in water (67 g/L) (25°C).

Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Chemical Identifiers

4349-07-9

Specifications

4349-07-9
MFCD02666079
Light sensitive
Soluble in water (67g/L) (25°C).
C1=C(C(=O)NC=N1)I
221.99
221.98
Solid
C4H3IN2O
1 g
5-iodopyrimidin-4-ol, 5-iodopyrimidin-4 3h-one, 4-hydroxy-5-iodopyrimidine, 5-iodo-1h-pyrimidin-4-one, 4 1h-pyrimidinone, 5-iodo, 5-iodopyrimidin-4 1h-one, 5-iodo-3h-pyrimidin-4-one, 5-iodo-3,4-dihydropyrimidin-4-one, 5-iodopyrimidin-4-ol;5-iodopyrimidin-4 1h-one
SMIGOGPUTQXOKF-UHFFFAOYSA-N
5-iodo-1H-pyrimidin-6-one
12356582
95%
4-Hydroxy-5-iodopyrimidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only