Learn More
4-Hydroxy-5-iodopyrimidine, 95%, Thermo Scientific Chemicals
Description
4-Hydroxy-5-iodopyrimidine is an intermediate used in pharmaceuticals and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Hydroxy-5-iodopyrimidine is an intermediate used in pharmaceuticals and chemical research.
Solubility
Soluble in water (67 g/L) (25°C).
Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Specifications
| Molecular Formula | C4H3IN2O |
| Quantity | 1 g |
| MDL Number | MFCD02666079 |
| Sensitivity | Light sensitive |
| Synonym | 5-iodopyrimidin-4-ol, 5-iodopyrimidin-4 3h-one, 4-hydroxy-5-iodopyrimidine, 5-iodo-1h-pyrimidin-4-one, 4 1h-pyrimidinone, 5-iodo, 5-iodopyrimidin-4 1h-one, 5-iodo-3h-pyrimidin-4-one, 5-iodo-3,4-dihydropyrimidin-4-one, 5-iodopyrimidin-4-ol;5-iodopyrimidin-4 1h-one |
| Solubility Information | Soluble in water (67g/L) (25°C). |
| InChI Key | SMIGOGPUTQXOKF-UHFFFAOYSA-N |
| SMILES | C1=C(C(=O)NC=N1)I |
| IUPAC Name | 5-iodo-1H-pyrimidin-6-one |
| Molecular Weight (g/mol) | 221.99 |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only