missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-4-phenylpiperidine 98.0+%, TCI America™

Catalog No. p-7130810
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
H09255G 5 g
H092525G 25 g
2 options

Catalog No. H09255G

Supplier: TCI America H09255G

Only null left

Chemical Identifiers

CAS 40807-61-2
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD00006000
InChI Key KQKFQBTWXOGINC-UHFFFAOYSA-N
Synonym 4-hydroxy-4-phenylpiperidine, 4-phenyl-4-piperidinol, 4-piperidinol, 4-phenyl, 4-phenyl-piperidin-4-ol, 4phenyl-4-piperidinol, 4-phenyl-4-piperidol, acmc-1amcr, 4-piperidinol,4-phenyl, 4-phenyl-piperidine-4-ol, 4-hydroxy-4phenylpiperidine
PubChem CID 96387
IUPAC Name 4-phenylpiperidin-4-ol
SMILES C1CNCCC1(C2=CC=CC=C2)O

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 5 g
Formula Weight 177.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-4-phenylpiperidine