missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-4-phenylpiperidine 98.0+%, TCI America™

Catalog No. H09255G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09255G 5 g
H092525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09255G Supplier TCI America Supplier No. H09255G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40807-61-2
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD00006000
InChI Key KQKFQBTWXOGINC-UHFFFAOYSA-N
Synonym 4-hydroxy-4-phenylpiperidine, 4-phenyl-4-piperidinol, 4-piperidinol, 4-phenyl, 4-phenyl-piperidin-4-ol, 4phenyl-4-piperidinol, 4-phenyl-4-piperidol, acmc-1amcr, 4-piperidinol,4-phenyl, 4-phenyl-piperidine-4-ol, 4-hydroxy-4phenylpiperidine
PubChem CID 96387
IUPAC Name 4-phenylpiperidin-4-ol
SMILES C1CNCCC1(C2=CC=CC=C2)O

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 5 g
Formula Weight 177.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-4-phenylpiperidine