missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-(trifluoromethoxy)benzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog No. p-7031505
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3260203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3260203 Supplier Thermo Scientific Chemicals Supplier No. H3260203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 53104-95-3 | Molecular Formula: C8H5F3O3 | Molecular Weight: 206.12 g/mol

4-Hydroxy-3-(trifluoromethoxy)benzaldehyde is a fluorinated compound, which is used for biochemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-3-(trifluoromethoxy)benzaldehyde is a fluorinated compound, which is used for biochemical research.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 53104-95-3
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
MDL Number MFCD04973780
InChI Key GPJSLRQIOKFRFL-UHFFFAOYSA-N
Synonym 4-hydroxy-3-trifluoromethoxy benzaldehyde, benzaldehyde, 4-hydroxy-3-trifluoromethoxy, 4-hydroxy-3-trifluoromethoxy benzaldehyde, 98+%, 4-oxidanyl-3-trifluoromethyloxy benzaldehyde
PubChem CID 7018050
IUPAC Name 4-hydroxy-3-(trifluoromethoxy)benzaldehyde
SMILES OC1=C(OC(F)(F)F)C=C(C=O)C=C1

Specifications

Melting Point 76°C to 80°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 206.12
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-3-(trifluoromethoxy)benzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only