missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-nitropyridine, 95%

Catalog No. AAB2096322
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2096322 100 g
AAB2096306 5 g
AAB2096314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2096322 Supplier Thermo Scientific Chemicals Supplier No. B2096322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5435-54-1 | C5H4N2O3 | 140.098 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5435-54-1
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.098
MDL Number MFCD00128874
InChI Key YUWOLBZMQDGRFV-UHFFFAOYSA-N
Synonym 4-hydroxy-3-nitropyridine, 3-nitropyridin-4-ol, 3-nitro-4-pyridinol, 3-nitro-4-hydroxypyridine, 3-nitropyridin-4 1h-one, 3-nitro-4-pyridone, 4-pyridinol, 3-nitro, 4 1h-pyridinone, 3-nitro, 3-nitro-1,4-dihydropyridin-4-one, 4-hydroxy-3-nitropytidine
PubChem CID 228506
IUPAC Name 3-nitro-1H-pyridin-4-one
SMILES C1=CNC=C(C1=O)[N+](=O)[O-]

Specifications

Melting Point ∼280°C (decomposition)
Quantity 100 g
Beilstein 1072928
Formula Weight 140.1
Percent Purity 95%
Chemical Name or Material 4-Hydroxy-3-nitropyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 611-127-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only