missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxy-3'-methylacetophenone 98.0+%, TCI America™

Catalog No. p-7130725
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07915G 5 g
H079125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07915G Supplier TCI America Supplier No. H07915G
Only null left

Chemical Identifiers

CAS 876-02-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00002231
InChI Key LXBHHIZIQVZGFN-UHFFFAOYSA-N
Synonym 4'-hydroxy-3'-methylacetophenone, 4-hydroxy-3-methylacetophenone, 1-4-hydroxy-3-methylphenyl ethanone, 1-4-hydroxy-3-methylphenyl ethan-1-one, 4-acetyl-2-methylphenol, ethanone, 1-4-hydroxy-3-methylphenyl, 4-hydroxy-3-methyl acetophenone, 1-4-hydroxy-3-methyl-phenyl-ethanone, 1-acetyl-4-hydroxy-3-methylbenzene
PubChem CID 70135
IUPAC Name 1-(4-hydroxy-3-methylphenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(O)C(C)=C1

Specifications

Melting Point 108°C
Quantity 5 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Hydroxy-3′-methylacetophenone