missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxy-3'-methylacetophenone, 98%

Catalog No. AAA1854003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1854003 1 g
AAA1854006 5 g
AAA1854014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1854003 Supplier Thermo Scientific Chemicals Supplier No. A1854003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 876-02-8 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 876-02-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00002231
InChI Key LXBHHIZIQVZGFN-UHFFFAOYSA-N
Synonym 4'-hydroxy-3'-methylacetophenone, 4-hydroxy-3-methylacetophenone, 1-4-hydroxy-3-methylphenyl ethanone, 1-4-hydroxy-3-methylphenyl ethan-1-one, 4-acetyl-2-methylphenol, ethanone, 1-4-hydroxy-3-methylphenyl, 4-hydroxy-3-methyl acetophenone, 1-4-hydroxy-3-methyl-phenyl-ethanone, 1-acetyl-4-hydroxy-3-methylbenzene
PubChem CID 70135
SMILES CC(=O)C1=CC=C(O)C(C)=C1

Specifications

Melting Point 107°C to 109°C
Boiling Point 175°C (1 mmHg)
Quantity 1 g
Beilstein 2041839
IUPAC Name 1-(4-hydroxy-3-methylphenyl)ethanone
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 4'-Hydroxy-3'-methylacetophenone
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 212-880-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only