missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Hydroxy-3'-methoxyacetophenone 98.0+%, TCI America™
Supplier: TCI America H026125G
Specifications
| 4′-Hydroxy-3′-methoxyacetophenone | |
| 115°C | |
| MFCD00008747 | |
| acetovanillone, apocynin, 4'-hydroxy-3'-methoxyacetophenone, 1-4-hydroxy-3-methoxyphenyl ethanone, acetoguaiacone, acetoguaiacon, apocynine, 4-acetyl-2-methoxyphenol, acetovanilone, acetovanyllon | |
| COC1=CC(=CC=C1O)C(C)=O | |
| 166.18 | |
| CHEBI:2781 | |
| ≥98.0% (GC) |
| 498-02-2 | |
| C9H10O3 | |
| 25 g | |
| DFYRUELUNQRZTB-UHFFFAOYSA-N | |
| 1-(4-hydroxy-3-methoxyphenyl)ethan-1-one | |
| 2214 | |
| 166.18 | |
| Crystalline Powder |
Chemical Identifiers
| 498-02-2 | |
| 166.18 | |
| DFYRUELUNQRZTB-UHFFFAOYSA-N | |
| 2214 | |
| 1-(4-hydroxy-3-methoxyphenyl)ethan-1-one |
| C9H10O3 | |
| MFCD00008747 | |
| acetovanillone, apocynin, 4'-hydroxy-3'-methoxyacetophenone, 1-4-hydroxy-3-methoxyphenyl ethanone, acetoguaiacone, acetoguaiacon, apocynine, 4-acetyl-2-methoxyphenol, acetovanilone, acetovanyllon | |
| CHEBI:2781 | |
| COC1=CC(=CC=C1O)C(C)=O |
Safety and Handling
EINECSNumber : (3)-0645
RTECSNumber : AM8800000
TSCA : Yes