missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxy-3'-methoxyacetophenone, 98%

Catalog No. AAA1043918
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1043918 50 g
AAA1043930 250 g
AAA1043936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1043918 Supplier Thermo Scientific Chemicals Supplier No. A1043918
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 498-02-2 | C9H10O3 | 166.18 g/mol

4'-Hydroxy-3'-methoxyacetophenone is used as an inhibitor of nicotinamide adenine dinucleotide phosphate (NADPH) oxidase, which is an enzyme responsible for reactive oxygen species production. It is also useful in the treatment of various inflammatory diseases and atherosclerosis. Further, it shows an anti-inflammatory activity and improves endothelial function by reducing oxidative stress. In addition to this, it is used for studying nitric oxide regulated processes in plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in ethanol, chloroform, ether, hot water, benzene, ethyl acetate and dimethyl sulfoxide. Insoluble in petroleum ether.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 498-02-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00008747
InChI Key DFYRUELUNQRZTB-UHFFFAOYSA-N
Synonym acetovanillone, apocynin, 4'-hydroxy-3'-methoxyacetophenone, 1-4-hydroxy-3-methoxyphenyl ethanone, acetoguaiacone, acetoguaiacon, apocynine, 4-acetyl-2-methoxyphenol, acetovanilone, acetovanyllon
PubChem CID 2214
ChEBI CHEBI:2781
IUPAC Name 1-(4-hydroxy-3-methoxyphenyl)ethanone
SMILES COC1=CC(=CC=C1O)C(C)=O

Specifications

Melting Point 112°C to 116°C
Boiling Point 295°C to 300°C
Quantity 50 g
Beilstein 637373
Merck Index 14,741
Solubility Information Soluble in ethanol,chloroform,ether,hot water,benzene,ethyl acetate and dimethyl sulfoxide. Insoluble in petroleum ether.
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 4'-Hydroxy-3'-methoxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-854-5
RTECSNumber AM8800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only