missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,6-dimethyl-2-pyrone, 97%

Catalog No. AAH2607406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2607406 5 g
AAH2607403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2607406 Supplier Thermo Scientific Chemicals Supplier No. H2607406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5192-62-1 | C7H8O3 | 140.138 g/mol

4-Hydroxy-3,6-dimethyl-2-pyrone is the reactant in the formation of unnatural novel polyketides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-3,6-dimethyl-2-pyrone is the reactant in the formation of unnatural novel polyketides.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 5192-62-1
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00090424
InChI Key VVBIGJOVPZMWGU-UHFFFAOYSA-N
Synonym 4-hydroxy-3,6-dimethyl-2h-pyran-2-one, 3,6-dimethyl-4-hydroxy-2-pyrone, chembl51918, 2h-pyran-2-one, 4-hydroxy-3,6-dimethyl, 2-hydroxy-3,6-dimethylpyran-4-one, hdmpo, 2-hydroxy-3,6-dimethyl-4h-pyran-4-one, acmc-20amh7, methyl triacetic lactone, 4-hydroxy-3,6-dimethyl-2-pyrone
PubChem CID 54690337
IUPAC Name 4-hydroxy-3,6-dimethylpyran-2-one
SMILES CC1=CC(=C(C(=O)O1)C)O

Specifications

Melting Point 209°C to 211°C
Quantity 5 g
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-3,6-dimethyl-2-pyrone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only