missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-3,6-dimethyl-2-pyrone, 97%
Description
4-Hydroxy-3,6-dimethyl-2-pyrone is the reactant in the formation of unnatural novel polyketides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Hydroxy-3,6-dimethyl-2-pyrone is the reactant in the formation of unnatural novel polyketides.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
4-Hydroxy-3,6-dimethyl-2-pyrone is the reactant in the formation of unnatural novel polyketides.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 209°C to 211°C |
| Quantity | 1 g |
| Formula Weight | 140.14 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Hydroxy-3,6-dimethyl-2-pyrone |
Safety and Handling
| missing translation for 'healthHazard1' | H315-H319-H335 |
| missing translation for 'healthHazard2' | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| missing translation for 'healthHazard3' | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only