missing translation for 'onlineSavingsMsg'
Learn More

1,4-Di(2-thienyl)-1,3-butadiene (mixture of isomers) 97.0+%, TCI America™

Catalog No. d20775g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20775G 5 g
D20771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20775G Supplier TCI America Supplier No. D20775G

Chemical Identifiers

CAS 23354-93-0
Molecular Formula C12H10S2
Molecular Weight (g/mol) 218.332
MDL Number MFCD00142549
InChI Key SEIGKYBFMXSLAH-UHFFFAOYSA-N
PubChem CID 53413352
IUPAC Name 2-(4-thiophen-2-ylbuta-1,3-dienyl)thiophene
SMILES C1=CSC(=C1)C=CC=CC2=CC=CS2

Specifications

Melting Point 171°C
Quantity 5 g
Formula Weight 218.33
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Di(2-thienyl)-1,3-butadiene (mixture of isomers)