missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-dimethylbenzaldehyde, 97+%

Catalog No. p-7028013
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2512303 1 g
AAB2512306 5 g
AAB2512314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2512303 Supplier Thermo Scientific Chemicals Supplier No. B2512303
Only null left

CAS: 2233-18-3 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2233-18-3
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00006946
InChI Key UYGBSRJODQHNLQ-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-hydroxybenzaldehyde, benzaldehyde, 4-hydroxy-3,5-dimethyl, 4-formyl-2,6-xylenol, 4-hydroxy-3,5-dimethyl-benzaldehyde, unii-23pa2pmp9f, 23pa2pmp9f, syringnldehyde, zlchem 510, pubchem8252, buttpark 9657-68
PubChem CID 75222
IUPAC Name 4-hydroxy-3,5-dimethylbenzaldehyde
SMILES CC1=CC(C=O)=CC(C)=C1O

Specifications

Melting Point 112°C to 116°C
Quantity 1 g
Beilstein 1908717
Sensitivity Air Sensitive
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3,5-dimethylbenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-774-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only