missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-dimethoxycinnamic acid, 98%

Catalog No. AAA1567614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1567614 25 g
AAA1567603 1 g
AAA1567606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1567614 Supplier Thermo Scientific Chemicals Supplier No. A1567614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 530-59-6 | C11H12O5 | 224.212 g/mol

4-Hydroxy-3,5-dimethoxycinnamic acid is a commonly used matrix in MALDI mass spectrometry. It is a useful matrix for a wide variety of peptides and proteins. It serves well as a matrix for MALDI due to its ability to absorb laser radiation and to also donate protons (H+) to the analyte of interest.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-3,5-dimethoxycinnamic acid is a commonly used matrix in MALDI mass spectrometry. It is a useful matrix for a wide variety of peptides and proteins. It serves well as a matrix for MALDI due to its ability to absorb laser radiation and to also donate protons (H+) to the analyte of interest.

Solubility
Soluble in methanol, ethanol and dimethyl sulfoxide.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 530-59-6
Molecular Formula C11H12O5
Molecular Weight (g/mol) 224.212
MDL Number MFCD00004401
InChI Key PCMORTLOPMLEFB-ONEGZZNKSA-N
Synonym sinapic acid, sinapinic acid, 3,5-dimethoxy-4-hydroxycinnamic acid, sinapinate, sinapate, trans-sinapic acid, 4-hydroxy-3,5-dimethoxycinnamic acid, synapoic acid, 2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid, unii-p0i60993ec
PubChem CID 637775
ChEBI CHEBI:15714
IUPAC Name (E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid
SMILES COC1=CC(=CC(=C1O)OC)C=CC(=O)O

Specifications

Melting Point ∼195°C (decomposition)
Quantity 25 g
Beilstein 2130006
Solubility Information Soluble in methanol,ethanol and dimethyl sulfoxide.
Formula Weight 224.21
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3,5-dimethoxycinnamic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-487-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only