missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxy-3',5'-dimethoxyacetophenone 98.0+%, TCI America™

Catalog No. p-7127434
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26665G 5 g
D266625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26665G Supplier TCI America Supplier No. D26665G
Only null left

Chemical Identifiers

CAS 2478-38-8
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00008748
InChI Key OJOBTAOGJIWAGB-UHFFFAOYSA-N
Synonym acetosyringone, 3',5'-dimethoxy-4'-hydroxyacetophenone, 1-4-hydroxy-3,5-dimethoxyphenyl ethanone, 4'-hydroxy-3',5'-dimethoxyacetophenone, 3,5-dimethoxy-4-hydroxyacetophenone, acetosyringon, ethanone, 1-4-hydroxy-3,5-dimethoxyphenyl, 1-4-hydroxy-3,5-dimethoxyphenyl ethan-1-one, acetosyringenin, acetophenone, 4'-hydroxy-3',5'-dimethoxy
PubChem CID 17198
ChEBI CHEBI:2404
IUPAC Name 1-(4-hydroxy-3,5-dimethoxyphenyl)ethan-1-one
SMILES COC1=CC(=CC(OC)=C1O)C(C)=O

Specifications

Melting Point 126°C
Quantity 5 g
Formula Weight 196.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Hydroxy-3′,5′-dimethoxyacetophenone
RTECSNumber AM8440000
TSCA No