missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxy-3',5'-dimethoxyacetophenone, 97%

Catalog No. AAB2263406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2263406 5 g
AAB2263403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2263406 Supplier Thermo Scientific Chemicals Supplier No. B2263406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2478-38-8 | C10H12O4 | 196.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2478-38-8
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00008748
InChI Key OJOBTAOGJIWAGB-UHFFFAOYSA-N
Synonym acetosyringone, 3',5'-dimethoxy-4'-hydroxyacetophenone, 1-4-hydroxy-3,5-dimethoxyphenyl ethanone, 4'-hydroxy-3',5'-dimethoxyacetophenone, 3,5-dimethoxy-4-hydroxyacetophenone, acetosyringon, ethanone, 1-4-hydroxy-3,5-dimethoxyphenyl, 1-4-hydroxy-3,5-dimethoxyphenyl ethan-1-one, acetosyringenin, acetophenone, 4'-hydroxy-3',5'-dimethoxy
PubChem CID 17198
ChEBI CHEBI:2404
IUPAC Name 1-(4-hydroxy-3,5-dimethoxyphenyl)ethanone
SMILES COC1=CC(=CC(OC)=C1O)C(C)=O

Specifications

Melting Point 117°C to 122°C
Quantity 5 g
Beilstein 1966119
Formula Weight 196.2
Percent Purity 97%
Chemical Name or Material 4'-Hydroxy-3',5'-dimethoxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-610-5
RTECSNumber AM8440000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only