missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-diiodobenzaldehyde 97.0+%, TCI America™

Catalog No. p-7131098
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15395G 5 g
H153925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15395G Supplier TCI America Supplier No. H15395G
Only null left

Chemical Identifiers

CAS 1948-40-9
Molecular Formula C7H4I2O2
Molecular Weight (g/mol) 373.92
MDL Number MFCD00014670
InChI Key WHLUEIMENHLCMY-UHFFFAOYSA-N
PubChem CID 74760
IUPAC Name 4-hydroxy-3,5-diiodobenzaldehyde
SMILES OC1=C(I)C=C(C=O)C=C1I

Specifications

Melting Point 204°C
Quantity 5 g
Synonym 3,5-diiodo-4-hydroxybenzaldehyde, benzaldehyde, 4-hydroxy-3,5-diiodo, 2,6-diiodo-4-formylphenol, 4-hydroxy-3,5-diiodo-benzaldehyde, acmc-1c01x, #, benzaldehyde,4-hydroxy-3,5-diiodo
Formula Weight 373.92
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-3,5-diiodobenzaldehyde