missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-diiodobenzaldehyde, 98+%

Catalog No. p-7043506
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0106009 10 g
AAL0106018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0106009 Supplier Thermo Scientific Chemicals Supplier No. L0106009
Only null left

CAS: 1948-40-9 | C7H4I2O2 | 373.92 g/mol

4-Hydroxy-3,5-diiodobenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-3,5-diiodobenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water. Sparingly soluble in chloroform and methanol.

Notes
Air & Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 1948-40-9
Molecular Formula C7H4I2O2
Molecular Weight (g/mol) 373.92
MDL Number MFCD00014670
InChI Key WHLUEIMENHLCMY-UHFFFAOYSA-N
Synonym 3,5-diiodo-4-hydroxybenzaldehyde, benzaldehyde, 4-hydroxy-3,5-diiodo, 2,6-diiodo-4-formylphenol, 4-hydroxy-3,5-diiodo-benzaldehyde, acmc-1c01x, #, benzaldehyde,4-hydroxy-3,5-diiodo
PubChem CID 74760
IUPAC Name 4-hydroxy-3,5-diiodobenzaldehyde
SMILES OC1=C(I)C=C(C=O)C=C1I

Specifications

Melting Point 198°C to 206°C
Quantity 10 g
Beilstein 1283414
Sensitivity Air and light sensitive
Solubility Information Insoluble in water. Sparingly soluble in chloroform and methanol.
Formula Weight 373.92
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-3,5-diiodobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-754-3
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only