missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-(trifluoromethyl)pyridine 98.0+%, TCI America™

Catalog No. H15285G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15285G 5 g
H15281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15285G Supplier TCI America Supplier No. H15285G

Chemical Identifiers

CAS 170886-13-2
Molecular Formula C6H4F3NO
Molecular Weight (g/mol) 163.099
MDL Number MFCD01571202
InChI Key LKHCWMQMRXAPHX-UHFFFAOYSA-N
Synonym 2-trifluoromethyl pyridin-4-ol, 4-hydroxy-2-trifluoromethyl pyridine, 4-hydroxy-2-trifluoromethylpyridine, 2-trifluoromethyl-4-hydroxypyridine, 2-trifluoromethyl-4-pyridinol, 4-pyridinol, 2-trifluoromethyl, 2-trifluoromethyl-1h-pyridin-4-one, 2-trifluoromethylpyridin-4-ol, 2-trifluoromethyl-1,4-dihydropyridin-4-one, pubchem17703
PubChem CID 10953849
IUPAC Name 2-(trifluoromethyl)-1H-pyridin-4-one
SMILES C1=CNC(=CC1=O)C(F)(F)F

Specifications

Melting Point 122°C
Color White-Yellow
Quantity 5 g
Formula Weight 163.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-2-(trifluoromethyl)pyridine