missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Hydroxy-2'-methylacetophenone 98.0+%, TCI America™
$171.34 - $550.61
Chemical Identifiers
| CAS | 875-59-2 |
|---|---|
| Molecular Formula | C9H10O2 |
| Molecular Weight (g/mol) | 150.18 |
| MDL Number | MFCD00002303 |
| InChI Key | IAMNVCJECQWBLZ-UHFFFAOYSA-N |
| Synonym | 4'-hydroxy-2'-methylacetophenone, 1-4-hydroxy-2-methylphenyl ethanone, 4-hydroxy-2-methylacetophenone, 2-methyl-4-hydroxyacetophenone, ethanone, 1-4-hydroxy-2-methylphenyl, 3-methyl-4-acetylphenol, 1-4-hydroxy-2-methylphenyl ethan-1-one, 1-acetyl-4-hydroxy-2-methylbenzene, ethanone,1-4-hydroxy-2-methylphenyl, 2'-methyl-4'-hydroxyacetophenone |
| PubChem CID | 70133 |
| ChEBI | CHEBI:87314 |
| IUPAC Name | 1-(4-hydroxy-2-methylphenyl)ethan-1-one |
| SMILES | CC(=O)C1=CC=C(O)C=C1C |
Chemical Identifiers
| 875-59-2 | |
| 150.18 | |
| IAMNVCJECQWBLZ-UHFFFAOYSA-N | |
| 70133 | |
| 1-(4-hydroxy-2-methylphenyl)ethan-1-one |
| C9H10O2 | |
| MFCD00002303 | |
| 4'-hydroxy-2'-methylacetophenone, 1-4-hydroxy-2-methylphenyl ethanone, 4-hydroxy-2-methylacetophenone, 2-methyl-4-hydroxyacetophenone, ethanone, 1-4-hydroxy-2-methylphenyl, 3-methyl-4-acetylphenol, 1-4-hydroxy-2-methylphenyl ethan-1-one, 1-acetyl-4-hydroxy-2-methylbenzene, ethanone,1-4-hydroxy-2-methylphenyl, 2'-methyl-4'-hydroxyacetophenone | |
| CHEBI:87314 | |
| CC(=O)C1=CC=C(O)C=C1C |
Specifications
| 875-59-2 | |
| White-Yellow | |
| MFCD00002303 | |
| 4'-hydroxy-2'-methylacetophenone, 1-4-hydroxy-2-methylphenyl ethanone, 4-hydroxy-2-methylacetophenone, 2-methyl-4-hydroxyacetophenone, ethanone, 1-4-hydroxy-2-methylphenyl, 3-methyl-4-acetylphenol, 1-4-hydroxy-2-methylphenyl ethan-1-one, 1-acetyl-4-hydroxy-2-methylbenzene, ethanone,1-4-hydroxy-2-methylphenyl, 2'-methyl-4'-hydroxyacetophenone | |
| CC(=O)C1=CC=C(O)C=C1C | |
| 150.18 | |
| CHEBI:87314 | |
| ≥98.0% (GC) | |
| 4′-Hydroxy-2′-methylacetophenone |
| 130°C | |
| C9H10O2 | |
| 5 g | |
| IAMNVCJECQWBLZ-UHFFFAOYSA-N | |
| 1-(4-hydroxy-2-methylphenyl)ethan-1-one | |
| 70133 | |
| 150.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No