missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxy-2'-methylacetophenone 98.0+%, TCI America™

Catalog No. H11245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11245G 5 g
H112425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H11245G Supplier TCI America Supplier No. H11245G

Chemical Identifiers

CAS 875-59-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00002303
InChI Key IAMNVCJECQWBLZ-UHFFFAOYSA-N
Synonym 4'-hydroxy-2'-methylacetophenone, 1-4-hydroxy-2-methylphenyl ethanone, 4-hydroxy-2-methylacetophenone, 2-methyl-4-hydroxyacetophenone, ethanone, 1-4-hydroxy-2-methylphenyl, 3-methyl-4-acetylphenol, 1-4-hydroxy-2-methylphenyl ethan-1-one, 1-acetyl-4-hydroxy-2-methylbenzene, ethanone,1-4-hydroxy-2-methylphenyl, 2'-methyl-4'-hydroxyacetophenone
PubChem CID 70133
ChEBI CHEBI:87314
IUPAC Name 1-(4-hydroxy-2-methylphenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(O)C=C1C

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 5 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Hydroxy-2′-methylacetophenone