missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxy-2'-methylacetophenone, 97%, Thermo Scientific Chemicals

Catalog No. p-3778798
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC121390250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC121390250 Supplier Thermo Scientific Chemicals Supplier No. 121390250

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 875-59-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00002303
InChI Key IAMNVCJECQWBLZ-UHFFFAOYSA-N
PubChem CID 70133
ChEBI CHEBI:87314
IUPAC Name 1-(4-hydroxy-2-methylphenyl)ethanone
SMILES CC(=O)C1=CC=C(O)C=C1C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 127°C to 132°C
Color Tan to White
Boiling Point 313°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC6H3(CH3)COCH3
Quantity 25 g
Beilstein 08, 111
Synonym 4'-hydroxy-2'-methylacetophenone, 1-4-hydroxy-2-methylphenyl ethanone, 4-hydroxy-2-methylacetophenone, 2-methyl-4-hydroxyacetophenone, ethanone, 1-4-hydroxy-2-methylphenyl, 3-methyl-4-acetylphenol, 1-4-hydroxy-2-methylphenyl ethan-1-one, 1-acetyl-4-hydroxy-2-methylbenzene, ethanone,1-4-hydroxy-2-methylphenyl, 2'-methyl-4'-hydroxyacetophenone
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 150.18
Percent Purity 97%
Physical Form Crystals or Powder
Chemical Name or Material 4'-Hydroxy-2'-methylacetophenone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 212-874-2

RUO – Research Use Only