missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-butanone 95.0+%, TCI America™

Catalog No. p-7130618
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
H058425ML 25 mL
H0584100ML 100 mL
H0584500ML 500 mL
3 options

Catalog No. H058425ML

Supplier: TCI America H058425ML

Only null left

Chemical Identifiers

CAS 590-90-9
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00059005
InChI Key LVSQXDHWDCMMRJ-UHFFFAOYSA-N
Synonym 4-hydroxy-2-butanone, 2-butanone, 4-hydroxy, methylolacetone, 3-oxo-1-butanol, 3-oxobutanol, 3-ketobutan-1-ol, monomethylolacetone, 4-butanol-2-one, unii-tcm0bj44mf, 2-hydroxyethyl methyl ketone
PubChem CID 111509
ChEBI CHEBI:41268
IUPAC Name 4-hydroxybutan-2-one
SMILES CC(=O)CCO

Specifications

Color Colorless
Boiling Point 170°C
Quantity 25 mL
Formula Weight 88.11
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Hydroxy-2-butanone
EINECSNumber (2)-3210
TSCA Yes