missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-butanone 95.0+%, TCI America™

Catalog No. H0584500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0584500ML 500 mL
H058425ML 25 mL
H0584100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0584500ML Supplier TCI America Supplier No. H0584500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 590-90-9
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00059005
InChI Key LVSQXDHWDCMMRJ-UHFFFAOYSA-N
Synonym 4-hydroxy-2-butanone, 2-butanone, 4-hydroxy, methylolacetone, 3-oxo-1-butanol, 3-oxobutanol, 3-ketobutan-1-ol, monomethylolacetone, 4-butanol-2-one, unii-tcm0bj44mf, 2-hydroxyethyl methyl ketone
PubChem CID 111509
ChEBI CHEBI:41268
IUPAC Name 4-hydroxybutan-2-one
SMILES CC(=O)CCO

Specifications

Color Colorless
Boiling Point 170°C
Quantity 500 mL
Formula Weight 88.11
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Hydroxy-2-butanone
EINECSNumber (2)-3210
TSCA Yes