missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-butanone, 95%

Catalog No. AAL1145622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1145622 100 g
AAL1145614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1145622 Supplier Thermo Scientific Chemicals Supplier No. L1145622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 590-90-9 | C4H8O2 | 88.106 g/mol

4-Hydroxy-2-butanone is used in the preparation of fused benzazepine molecules as selective D3 receptor antagonists with pharmaceutical activity. It is also used in the preparation of verrucarin and mevalonic acid lactone. Further, it is used to prepare 3-buten-2-one through dehydration over an anatase titanium dioxide catalyst. It is also employed in the preparation of 4-methyl-5-hydroxymethylthiazole, (±)-lineatin and beta-hydroxylactones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-2-butanone is used in the preparation of fused benzazepine molecules as selective D3 receptor antagonists with pharmaceutical activity. It is also used in the preparation of verrucarin and mevalonic acid lactone. Further, it is used to prepare 3-buten-2-one through dehydration over an anatase titanium dioxide catalyst. It is also employed in the preparation of 4-methyl-5-hydroxymethylthiazole, (±)-lineatin and beta-hydroxylactones.

Solubility
Miscible with water, alcohol, ethanol and ether.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 590-90-9
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00059005
InChI Key LVSQXDHWDCMMRJ-UHFFFAOYSA-N
Synonym 4-hydroxy-2-butanone, 2-butanone, 4-hydroxy, methylolacetone, 3-oxo-1-butanol, 3-oxobutanol, 3-ketobutan-1-ol, monomethylolacetone, 4-butanol-2-one, unii-tcm0bj44mf, 2-hydroxyethyl methyl ketone
PubChem CID 111509
ChEBI CHEBI:41268
IUPAC Name 4-hydroxybutan-2-one
SMILES CC(=O)CCO

Specifications

Density 1.02
Boiling Point 73°C to 76°C (12 mmHg)
Flash Point 89°C (192°F)
Refractive Index 1.431
Quantity 100 g
Beilstein 1737666
Solubility Information Miscible with water,alcohol,ethanol and ether.
Formula Weight 88.11
Percent Purity 95%
Chemical Name or Material 4-Hydroxy-2-butanone
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 209-693-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only