missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2,2,6,6-tetramethylpiperidine 98.0+%, TCI America™

Catalog No. T091025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T091025G 25 g
T0910500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T091025G Supplier TCI America Supplier No. T091025G

Chemical Identifiers

CAS 2403-88-5
Molecular Formula C9H19NO
Molecular Weight (g/mol) 157.257
MDL Number MFCD00005983
InChI Key VDVUCLWJZJHFAV-UHFFFAOYSA-N
Synonym 2,2,6,6-tetramethyl-4-piperidinol, lastar a, 4-piperidinol, 2,2,6,6-tetramethyl, 4-hydroxy-2,2,6,6-tetramethylpiperidine, 2,2,6,6-tetramethyl-4-hydroxypiperidine, unii-is23q8egz6, is23q8egz6, 2,2,6,6-tetramethyl-piperidin-4-ol, pubchem23863
PubChem CID 75471
IUPAC Name 2,2,6,6-tetramethylpiperidin-4-ol
SMILES CC1(CC(CC(N1)(C)C)O)C

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 25 g
UN Number 3259
Formula Weight 157.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-2,2,6,6-tetramethylpiperidine
EINECSNumber (5)-0776
RTECSNumber TN7871675
TSCA Yes