missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2,2,6,6-tetramethylpiperidine, 98%

Catalog No. AAB2266406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2266406 5 g
AAB2266414 25 g
AAB2266422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2266406 Supplier Thermo Scientific Chemicals Supplier No. B2266406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2403-88-5 | C9H19NO | 157.257 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2403-88-5
Molecular Formula C9H19NO
Molecular Weight (g/mol) 157.257
MDL Number MFCD00005983
InChI Key VDVUCLWJZJHFAV-UHFFFAOYSA-N
Synonym 2,2,6,6-tetramethyl-4-piperidinol, lastar a, 4-piperidinol, 2,2,6,6-tetramethyl, 4-hydroxy-2,2,6,6-tetramethylpiperidine, 2,2,6,6-tetramethyl-4-hydroxypiperidine, unii-is23q8egz6, is23q8egz6, 2,2,6,6-tetramethyl-piperidin-4-ol, pubchem23863
PubChem CID 75471
IUPAC Name 2,2,6,6-tetramethylpiperidin-4-ol
SMILES CC1(CC(CC(N1)(C)C)O)C

Specifications

Melting Point 129°C to 133°C
Density 1.085
pH 11.2
Boiling Point 212°C to 215°C
Flash Point 124°C (255°F)
Quantity 5 g
UN Number UN3259
Beilstein 105039
Formula Weight 157.26
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-2,2,6,6-tetramethylpiperidine
Show More Show Less
Hazard Category H312-H315-H319-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber 219-291-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only