missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-1-methylpiperidine 98.0+%, TCI America™

Catalog No. p-7132450
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
M093325G 25 g
M0933500G 500 g
2 options

Catalog No. M093325G

Supplier: TCI America M093325G

Only null left

Chemical Identifiers

CAS 106-52-5
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00006500
InChI Key BAUWRHPMUVYFOD-UHFFFAOYSA-N
Synonym 4-hydroxy-1-methylpiperidine, n-methyl-4-piperidinol, 1-methyl-4-piperidinol, 4-hydroxy-n-methylpiperidine, n-methyl-4-hydroxypiperidine, 4-piperidinol, 1-methyl, 1-methyl-4-hydroxypiperidine, n-methylpiperidol, n-methyl-4-hydroxy piperidine, 4-hydroxy-1-methyl-piperidine
PubChem CID 66048
IUPAC Name 1-methylpiperidin-4-ol
SMILES CN1CCC(CC1)O

Specifications

Boiling Point 105°C
Quantity 25 g
Formula Weight 115.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Hydroxy-1-methylpiperidine
EINECSNumber (9)-1463
TSCA Yes