missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-1-(2-hydroxyethyl)-2,2,6,6-tetramethylpiperidine 98.0+%, TCI America™
$328.08 - $733.13
Chemical Identifiers
| CAS | 52722-86-8 |
|---|---|
| Molecular Formula | C11H23NO2 |
| Molecular Weight (g/mol) | 201.31 |
| MDL Number | MFCD01321174 |
| InChI Key | STEYNUVPFMIUOY-UHFFFAOYSA-N |
| Synonym | 1-(2-Hydroxyethyl)-2,2,6,6-tetramethyl-4-piperidinol, 4-Hydroxy-2,2,6,6-tetramethyl-1-piperidineethanol |
| PubChem CID | 104301 |
| IUPAC Name | 1-(2-hydroxyethyl)-2,2,6,6-tetramethylpiperidin-4-ol |
| SMILES | CC1(CC(CC(N1CCO)(C)C)O)C |
Chemical Identifiers
| 52722-86-8 | |
| 201.31 | |
| STEYNUVPFMIUOY-UHFFFAOYSA-N | |
| 104301 | |
| CC1(CC(CC(N1CCO)(C)C)O)C |
| C11H23NO2 | |
| MFCD01321174 | |
| 1-(2-Hydroxyethyl)-2,2,6,6-tetramethyl-4-piperidinol, 4-Hydroxy-2,2,6,6-tetramethyl-1-piperidineethanol | |
| 1-(2-hydroxyethyl)-2,2,6,6-tetramethylpiperidin-4-ol |
Specifications
| 52722-86-8 | |
| White | |
| MFCD01321174 | |
| 1-(2-Hydroxyethyl)-2,2,6,6-tetramethyl-4-piperidinol, 4-Hydroxy-2,2,6,6-tetramethyl-1-piperidineethanol | |
| CC1(CC(CC(N1CCO)(C)C)O)C | |
| 201.31 | |
| 201.31 | |
| Crystalline Powder |
| 186°C | |
| C11H23NO2 | |
| 5 g | |
| STEYNUVPFMIUOY-UHFFFAOYSA-N | |
| 1-(2-hydroxyethyl)-2,2,6,6-tetramethylpiperidin-4-ol | |
| 104301 | |
| ≥98.0% (GC) | |
| 4-Hydroxy-1-(2-hydroxyethyl)-2,2,6,6-tetramethylpiperidine |
Safety and Handling
TSCA : Yes