missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-1,2,2,6,6-pentamethylpiperidine 98.0+%, TCI America™

Catalog No. p-7130888
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H106825G 25 g
H1068500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H106825G Supplier TCI America Supplier No. H106825G
Only null left

Chemical Identifiers

CAS 2403-89-6
Molecular Formula C10H21NO
Molecular Weight (g/mol) 171.284
InChI Key NWHNXXMYEICZAT-UHFFFAOYSA-N
Synonym 1,2,2,6,6-pentamethyl-4-piperidinol, 4-piperidinol, 1,2,2,6,6-pentamethyl, 4-hydroxy-1,2,2,6,6-pentamethylpiperidine, unii-lr520bam5v, 1,2,2,6,6-pentamethyl-4-hydroxypiperidine, lr520bam5v, methyl-taa-ol, hopemp, pubchem22251, acmc-209g8a
PubChem CID 75472
IUPAC Name 1,2,2,6,6-pentamethylpiperidin-4-ol
SMILES CC1(CC(CC(N1C)(C)C)O)C

Specifications

Melting Point 76°C
Color White
Boiling Point 120°C
Quantity 25 g
Formula Weight 171.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-1,2,2,6,6-pentamethylpiperidine
EINECSNumber (5)-6350
TSCA Yes