missing translation for 'onlineSavingsMsg'
Learn More

4-Hexylthiophene-2-carbonitrile 95.0+%, TCI America™

Catalog No. h1503200mg
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1503200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H1503200MG Supplier TCI America Supplier No. H1503200MG

Chemical Identifiers

CAS 1224430-39-0
Molecular Formula C11H15NS
Molecular Weight (g/mol) 193.308
MDL Number MFCD18413940
InChI Key LFLOKABSZNAYLC-UHFFFAOYSA-N
Synonym 2-Cyano-4-hexylthiophene
PubChem CID 58420906
IUPAC Name 4-hexylthiophene-2-carbonitrile
SMILES CCCCCCC1=CSC(=C1)C#N

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 193.31
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Hexylthiophene-2-carbonitrile