missing translation for 'onlineSavingsMsg'
Learn More

4-Hexylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. H14891G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H14891G Supplier TCI America Supplier No. H14891G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105365-50-2
Molecular Formula C12H19BO2
MDL Number MFCD04039029
InChI Key CFYBKGAFWFRFRJ-UHFFFAOYSA-N
PubChem CID 4197864
IUPAC Name (4-hexylphenyl)boronic acid

Specifications

Color White
Quantity 1 g
Synonym 4-hexylphenyl boronic acid, 4-n-hexylphenylboronic acid, 4-hex-1-yl benzeneboronic acid, 1-borono-4-hex-1-yl benzene, 4-n-hexylbenzeneboronic acid, boronic acid, 4-hexylphenyl, 4-hexylphenylboronicacid, p-hexylphenylboronic acid, 4-hexylbenzeneboronic acid, 4-hexylphenyl-boronic acid
SMILES B(C1=CC=C(C=C1)CCCCCC)(O)O
Molecular Weight (g/mol) 206.092
Formula Weight 206.09
Physical Form Crystalline Powder
Chemical Name or Material 4-Hexylphenylboronic Acid (contains varying amounts of Anhydride)