missing translation for 'onlineSavingsMsg'
Learn More

4'-Hexylacetophenone 96.0+%, TCI America™

Catalog No. H066925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H066925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H066925G Supplier TCI America Supplier No. H066925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 37592-72-6
Molecular Formula C14H20O
Molecular Weight (g/mol) 204.313
MDL Number MFCD00043677
InChI Key WWBVHJKFJZBRSO-UHFFFAOYSA-N
Synonym p-hexylacetophenone, 1-4-hexylphenyl ethanone, 4'-hexylacetophenone, 1-4-hexylphenyl ethan-1-one, 4'-n-hexylacetophenone, ethanone, 1-4-hexylphenyl, 4-n-hexylacetophenone, 4-hexylacetophenone, 1-acetyl-4-hexylbenzene, agarose gpg/le
PubChem CID 123462
IUPAC Name 1-(4-hexylphenyl)ethanone
SMILES CCCCCCC1=CC=C(C=C1)C(=O)C

Specifications

Color Yellow
Boiling Point 172°C
Quantity 25 g
Formula Weight 204.31
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Hexylacetophenone
EINECSNumber (3)-3540
TSCA Yes