missing translation for 'onlineSavingsMsg'
Learn More

4-Hexen-3-one, 98%, trans-isomer >95%

Catalog No. AAH2744509
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2744509 10 g
AAH2744518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2744509 Supplier Thermo Scientific Chemicals Supplier No. H2744509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2497-21-4 | C6H10O | 98.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2497-21-4
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00010631
InChI Key FEWIGMWODIRUJM-HYXAFXHYSA-N
Synonym 4-hexen-3-one, 2-hexen-4-one, 2-hexene-4-one, e-hex-4-en-3-one, 4-hexene-3-one, fema no. 3352, e-4-hexen-3-one, 4-hexen-3-one, predominantly trans, e-ch3ch=chc =o c2h5, trans-4-hexen-3-one
PubChem CID 5365811
IUPAC Name (4Z)-hex-4-en-3-one
SMILES CCC(=O)\C=C/C

Specifications

Density 0.858
Boiling Point 135°C to 137°C
Flash Point 34°C (93°F)
Odor Characteristic
Refractive Index 1.44
Quantity 10 g
UN Number UN1224
Formula Weight 98.15
Percent Purity 98%
Assay trans-isomer >95%
Chemical Name or Material 4-Hexen-3-one
Show More Show Less
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H226-H302
Flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 219-681-2
RTECSNumber MP8659900
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only