missing translation for 'onlineSavingsMsg'
Learn More

4-Heptyloxybenzaldehyde 98.0+%, TCI America™

Catalog No. H023825ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H023825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H023825ML Supplier TCI America Supplier No. H023825ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27893-41-0
Molecular Formula C14H20O2
Molecular Weight (g/mol) 220.312
MDL Number MFCD00016616
InChI Key YBCKMIZXHKVONZ-UHFFFAOYSA-N
Synonym 4-heptyloxybenzaldehyde, 4-heptyloxy benzaldehyde, p-heptyloxybenzaldehyde, 4-n-heptyloxybenzaldehyde, benzaldehyde, 4-heptyloxy, p-heptyloxy benzaldehyde, 4-n-heptoxybenzaldehyde, 4-heptyloxy-benzaldehyde, p-n-heptoxybenzaldehyde, acmc-209gzj
PubChem CID 119740
IUPAC Name 4-heptoxybenzaldehyde
SMILES CCCCCCCOC1=CC=C(C=C1)C=O

Specifications

Color Yellow
Boiling Point 164°C
Quantity 25 mL
Formula Weight 220.31
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Heptyloxybenzaldehyde